
6-Mercapto-1-hexanol

Synonym(s):
6-Hydroxy-1-hexanethiol, 6-MCH, 6-Mercaptohexan-1-ol, 6-Mercaptohexanol, MCH
Linear Formula:
HS(CH2)6OH
CAS Number:
Molecular Weight:
134.24
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing