
4-Hydroxymidazolam solution

Synonym(s):
4-Hydroxymidazolam solution
Empirical Formula (Hill Notation):
C18H13ClFN3O
CAS Number:
Molecular Weight:
341.77
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing