4-Amino-3-nitrophenol
Synonym(s):
4-Hydroxy-2-nitroaniline
Linear Formula:
H2NC6H3(NO2)OH
CAS Number:
Molecular Weight:
154.12
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing