Skip to Content
Merck
3-Bromo-2,6-dimethylpyridine 97%

3-Bromo-2,6-dimethylpyridine

3-Bromo-2,6-dimethylpyridine 97%
Empirical Formula (Hill Notation):
C7H8BrN
CAS Number:
Molecular Weight:
186.05

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing