Skip to Content
Merck
3,4-Dihydroxyphenylacetic acid analytical standard

3,4-Dihydroxyphenylacetic acid

3,4-Dihydroxyphenylacetic acid analytical standard
Synonym(s):
DOPAC, Homoprotocatechuic acid
Linear Formula:
(HO)2C6H3CH2CO2H
CAS Number:
Molecular Weight:
168.15
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing