
2-Chlorotritanol

Synonym(s):
(o-Chlorophenyl)diphenylmethanol
Empirical Formula (Hill Notation):
C19H15ClO
CAS Number:
Molecular Weight:
294.77
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing