Skip to Content
Merck
2,6-Dichlorodiphenylamine analytical standard

2,6-Dichlorodiphenylamine

2,6-Dichlorodiphenylamine analytical standard
Synonym(s):
N-Phenyl-2,6-dichloroaniline
Linear Formula:
Cl2C6H3NHC6H5
CAS Number:
Molecular Weight:
238.11
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing