
2,5-Dimethoxyphenol

Empirical Formula (Hill Notation):
C8H10O3
CAS Number:
Molecular Weight:
154.16
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing