Skip to Content
Merck
2,2,2-Trichloroethyl chloroformate purum, ≥98.0% (NT)

2,2,2-Trichloroethyl chloroformate

2,2,2-Trichloroethyl chloroformate purum, ≥98.0% (NT)
Synonym(s):
2,2,2-Trichloroethoxycarbonyl chloride
Linear Formula:
ClCOOCH2CCl3
CAS Number:
Molecular Weight:
211.86
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing