![(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol for Copper-free Click Chemistry](/deepweb/assets/sigmaaldrich/product/structures/171/632/0556139a-2db5-4678-a6ec-a26a693fd574/640/0556139a-2db5-4678-a6ec-a26a693fd574.png)
(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol
![(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol for Copper-free Click Chemistry](/deepweb/assets/sigmaaldrich/product/structures/171/632/0556139a-2db5-4678-a6ec-a26a693fd574/640/0556139a-2db5-4678-a6ec-a26a693fd574.png)
Synonym(s):
(1α,8α,9β)-Bicyclo[6.1.0]non-4-yne-9-methanol, endo-9-Hydroxymethylbicyclo[6.1.0]non-4-yne, BCN-OH
Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing