Z727938
Hettich® swing-out rotor, for plates
for UNIVERSAL 320/320R Centrifuge, Swing-out Rotor, for plates
Synonym(s):
Hettich® centrifuge rotors
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
Recommended Products
sterility
non-sterile
feature
to hold 4 x MTP or CP plates
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1645
parameter
1,896 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Universal 320/320R
Looking for similar products? Visit Product Comparison Guide
General description
Plates and plate carriers not included.
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
required but not provided
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service