SRP6315
Alpha 2 Macroglobulin, Fast Form from human plasma
≥95% (SDS-PAGE)
Synonym(s):
A2M, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5, FWP007
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
General description
α 2 Macroglobulin (A2M) is a plasma protease inhibitor which has been shown to exist in two forms. The Slow Form of A2M (S-A2M) is the form which possesses the ability to bind and inhibit proteases by a “trap” method. The Fast Form of A2M (F-A2M) is generated when S-A2M undergoes a conformational change due to either entrapment of a protease in the A2M bait region, or chemical cleavage of an internal thiol ester bond located near the bait region. F-A2M does not possess the ability to bind and inhibit protease activity. F-A2M is rapidly taken up by the liver, with a half-life of 2-4 minutes. In vivo, F-A2M typically represents only 0.17–0.7% of the total A2M in blood plasma of adults. The F-A2M plasma concentration is, however, increased in many disease states including pancreatitis, multiple sclerosis and sepsis. F-A2M has also been implicated in the inhibition of amyloid formation associated with Alzheimer′s disease and spongiform encephalopathy.
α 2 macroglobulin (A2M) is a major serum protein found at concentrations of 240 mg/100 mL in men and 290 mg/100 mL in women. It is produced by the liver and is a major component of the α-2 band in protein electrophoresis. It has a molecular weight of 800,000Da.
Biochem/physiol Actions
α 2 macroglobulin (A2M) functions as a broad-spectrum protease-binding protein. In addition, it acts as an irreversible inhibitor of a variety of proteinases. A2M expressed in the brain interacts with multiple extracellular ligands and is interiorized by neurons and astrocytes. In the brain of Alzheimer′s disease (AD) patients, A2M has been localized to diffuse amyloid plaques. A2M interacts with soluble β-amyloid and facilitates its degradation.
Physical form
Lyophilized from 100 mM Na Phosphate, pH 7.2.
Reconstitution
In water or aqueous buffer
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Interleukin-6 and α2-macroglobulin indicate an acute-phase state in Alzheimer's disease cortices.
Bauer, Joachim, et al.
Febs Letters, 285(1), 111-114 (1991)
The interaction of α2-macroglobulin with proteinases. Characteristics and specificity of the reaction, and a hypothesis concerning its molecular mechanism.
Barrett A J and Phyllis M S
The Biochemical Journal, 133(4), 709-724 (1973)
Loic Auderset et al.
Frontiers in cell and developmental biology, 8, 564351-564351 (2020-12-08)
Low-density lipoprotein receptor-related protein 1 (LRP1) is a large, endocytic cell surface receptor that is highly expressed by oligodendrocyte progenitor cells (OPCs) and LRP1 expression is rapidly downregulated as OPCs differentiate into oligodendrocytes (OLs). We report that the conditional deletion
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service