Z150851
Liquid nitrogen dipper
for removing small quantities of liquid nitrogen (LN2) from storage dewars
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
Recommended Products
General description
Safe and convenient LN2 dipper. The low conducting measuring rod will clearly show the LN2 level of a dewar in inches and centimeters.
Compatible with the following range of cryogenic storage dewars, LD series: LD5, LD10, LD25, Classic 25, LD35 and LD50
Compatible with the following range of cryogenic storage dewars, LD series: LD5, LD10, LD25, Classic 25, LD35 and LD50