Merck
  • A Complementary Scale of Biased Agonism for Agonists with Differing Maximal Responses.
A Complementary Scale of Biased Agonism for Agonists with Differing Maximal Responses.
Scientific reports (2017-11-15)
Javier Burgueño, Marta Pujol, Xavier Monroy, David Roche, Maria Jose Varela, Manuel Merlos, Jesús Giraldo
ABSTRACT
Compelling data in the literature from the recent years leave no doubt about the pluridimensional nature of G protein-coupled receptor function and the fact that some ligands can couple with different efficacies to the multiple pathways that a receptor can signal through, a phenomenon most commonly known as functional selectivity or biased agonism. Nowadays, transduction coefficients (log(τ/K
Sigma-Aldrich
Endomorphin II
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)