Merck
  • Synthesis, Crystal Structures and Catalytic Oxidation of Manganese(III) Complexes Derived from Salen-Type Schiff Base N,N'-Bis(5-nitrosalicylidene)ethane-1,2-diamine.
Synthesis, Crystal Structures and Catalytic Oxidation of Manganese(III) Complexes Derived from Salen-Type Schiff Base N,N'-Bis(5-nitrosalicylidene)ethane-1,2-diamine.
Acta chimica Slovenica (2017-06-18)
Qing-Bin Li, Yong-Jun Han, Gan-Qing Zhao, Ling-Wei Xue
ABSTRACT
Two mononuclear Schiff base manganese(III) complexes, [MnL(N3)(OH2)]·CH3OH (1) and [MnL(NCS)(OH2)] · H2O (2), where L is the dianionic form of N,N'-bis(5-nitrosalicylidene)ethane-1,2-diamine, have been prepared and characterized by elemental analysis, IR and UV-Vis spectroscopy and single crystal X-ray diffraction. The Mn atom in each complex is in an octahedral coordination. Molecules of the complexes are linked through intermolecular hydrogen bonds. Catalytic properties for epoxidation of styrene by the complexes using PhIO and NaOCl as oxidant have been studied.
Sigma-Aldrich
2-Hydroxy-5-nitrobenzaldehyde, 98%