Merck
  • Serological responses and biomarker evaluation in mice and pigs exposed to tsetse fly bites.
Serological responses and biomarker evaluation in mice and pigs exposed to tsetse fly bites.
PLoS neglected tropical diseases (2014-05-24)
Guy Caljon, Reta Duguma, Reginald De Deken, Stijn Schauvliege, Frank Gasthuys, Luc Duchateau, Jan Van Den Abbeele
ABSTRACT
Tsetse flies are obligate blood-feeding insects that transmit African trypanosomes responsible for human sleeping sickness and nagana in livestock. The tsetse salivary proteome contains a highly immunogenic family of the endonuclease-like Tsal proteins. In this study, a recombinant version of Tsal1 (rTsal1) was evaluated in an indirect ELISA to quantify the contact with total Glossina morsitans morsitans saliva, and thus the tsetse fly bite exposure. Mice and pigs were experimentally exposed to different G. m. morsitans exposure regimens, followed by a long-term follow-up of the specific antibody responses against total tsetse fly saliva and rTsal1. In mice, a single tsetse fly bite was sufficient to induce detectable IgG antibody responses with an estimated half-life of 36-40 days. Specific antibody responses could be detected for more than a year after initial exposure, and a single bite was sufficient to boost anti-saliva immunity. Also, plasmas collected from tsetse-exposed pigs displayed increased anti-rTsal1 and anti-saliva IgG levels that correlated with the exposure intensity. A strong correlation between the detection of anti-rTsal1 and anti-saliva responses was recorded. The ELISA test performance and intra-laboratory repeatability was adequate in the two tested animal models. Cross-reactivity of the mouse IgGs induced by exposure to different Glossina species (G. m. morsitans, G. pallidipes, G. palpalis gambiensis and G. fuscipes) and other hematophagous insects (Stomoxys calcitrans and Tabanus yao) was evaluated. This study illustrates the potential use of rTsal1 from G. m. morsitans as a sensitive biomarker of exposure to a broad range of Glossina species. We propose that the detection of anti-rTsal1 IgGs could be a promising serological indicator of tsetse fly presence that will be a valuable tool to monitor the impact of tsetse control efforts on the African continent.
Sigma-Aldrich
Anti-Pig IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.