Merck
  • Design of a flexible cell-based assay for the evaluation of heat shock protein 70 expression modulators.
Design of a flexible cell-based assay for the evaluation of heat shock protein 70 expression modulators.
Assay and drug development technologies (2010-12-08)
James H Ahn, Wenjie Luo, Joungnam Kim, Anna Rodina, Cristina C Clement, Julia Aguirre, Weilin Sun, Yanlong Kang, Ronnie Maharaj, Kamalika Moulick, Danuta Zatorska, Malgorzata Kokoszka, Jeffrey L Brodsky, Gabriela Chiosis
ABSTRACT
Heat shock protein 70 (Hsp70) is a chaperone protein that helps protect against cellular stress, a function that may be co-opted to fight human diseases. In particular, the upregulation of Hsp70 can suppress the neurotoxicity of misfolded proteins, suggesting possible therapeutic strategies in neurodegenerative diseases. Alternatively, in cancer cells where high levels of Hsp70 inhibit both intrinsic and extrinsic apoptotic pathways, a reduction in Hsp70 levels may induce apoptosis. To evaluate and identify, in a single assay format, small molecules that induce or inhibit endogenous Hsp70, we have designed and optimized a microtiter assay that relies on whole-cell immunodetection of Hsp70. The assay utilizes a minimal number of neuronal or cancer cells, yet is sufficiently sensitive and reproducible to permit quantitative determinations. We further validated the assay using a panel of Hsp70 modulators. In conclusion, we have developed an assay that is fast, robust, and cost efficient. As such, it can be implemented in most research laboratories. The assay should greatly improve the speed at which novel Hsp70 inducers and inhibitors of expression can be identified and evaluated.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
17-(Allylamino)-17-demethoxygeldanamycin, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Hsp70 Mouse mAb (C92F3A-5), liquid, clone C92F3A-5, Calbiochem®
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture