Merck
  • Nano- and microparticles as adjuvants in vaccine design: success and failure is related to host natural antibodies.
Nano- and microparticles as adjuvants in vaccine design: success and failure is related to host natural antibodies.
Vaccine (2006-07-18)
Michael S Sinyakov, Moti Dror, Tammy Lublin-Tennenbaum, Samuel Salzberg, Shlomo Margel, Ramy R Avtalion
ABSTRACT
Bovine serum albumin (BSA) and the surface A-layer protein (AP) of an atypical strain of fish bacterial pathogen Aeromonas salmonicida were covalently linked with polymeric nano- and microparticles, and antigenicity of the resulted conjugates was compared in mice and goldfish. Distinct albeit different levels of natural BSA and AP antibodies were present in both animal species. Significant stimulation of the anti-AP antibody response in mice strikingly contrasted to unresponsiveness or even suppression in fish. The results negatively correlate with the levels of respective natural antibodies in the host and are discussed in context of problems related to fish vaccination. The work reinforces the instructive role of natural antibodies in adaptive immune response.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Pricing and availability is not currently available.