Merck
Betulinic acid, a natural compound with potent anticancer effects.
Anti-cancer drugs (2010-01-16)
Franziska B Mullauer, Jan H Kessler, Jan Paul Medema
ABSTRACT
New therapies using novel mechanisms to induce tumor cell death are needed with plants playing a crucial role as a source for potential anticancer compounds. One highly promising class of natural compounds are the triterpenoids with betulinic acid (BetA) as the most prominent representative. In-vitro studies have identified this agent as potently effective against a wide variety of cancer cells, also those derived from therapy-resistant and refractory tumors, whereas it has been found to be relatively nontoxic for healthy cells. In-vivo preclinically applied BetA showed some remarkable anticancer effects and a complete absence of systemic toxicity in rodents. BetA also cooperated with other therapies to induce tumor cell death and several potent derivatives have been discovered. Its antitumor activity has been related to its direct effects on mitochondria where it induces Bax/Bak-independent cytochrome-c release.
Product Description
Supelco
Betulinic acid, analytical standard
Sigma-Aldrich
Betulinic acid, technical grade, 90%
Sigma-Aldrich
Betulinic acid, ≥98% (HPLC)