Merck
Fragrance material review on nerol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
A Lapczynski, R J Foxenberg, S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of nerol when used as a fragrance ingredient is presented.
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
Supelco
Nerol, analytical standard