Merck
  • Fluoro-substituted ketones from nitriles using acidic and basic reaction conditions.
Fluoro-substituted ketones from nitriles using acidic and basic reaction conditions.
Tetrahedron letters (2012-03-03)
Erum Raja, Douglas A Klumpp
ABSTRACT
Fluoro-substituted aliphatic nitriles are shown to undergo the Houben-Hoesch reactions with arenes in CF(3)SO(3)H to give fluoro-substituted ketones in good yields. The fluorine substituents appear to enhance the reactivities of the nitriles (and the nitrilium ion intermediates) compared to similar aliphatic nitriles. Fluoro-substituted ketones are also shown to be accessible through the reactions of organometallic reagents and fluoro-substituted nitriles.
Sigma-Aldrich
Fluoroacetonitrile, 98%
Pricing and availability is not currently available.