Merck
  • Application of qualitative and quantitative real-time PCR, direct sequencing, and terminal restriction fragment length polymorphism analysis for detection and identification of polymicrobial 16S rRNA genes in ascites.
Application of qualitative and quantitative real-time PCR, direct sequencing, and terminal restriction fragment length polymorphism analysis for detection and identification of polymicrobial 16S rRNA genes in ascites.
Journal of clinical microbiology (2014-03-14)
Sandra Krohn, Stephan Böhm, Cornelius Engelmann, Jan Hartmann, Annika Brodzinski, Antonis Chatzinotas, Katharina Zeller, Delia Prywerek, Ingo Fetzer, Thomas Berg
ABSTRACT
Qualitative and quantitative 16S rRNA gene-based real-time PCR and direct sequencing were applied for rapid detection and identification of bacterial DNA (bactDNA) in 356 ascites samples. bactDNA was detected in 35% of samples, with a mean of 3.24 log copies ml(-1). Direct sequencing of PCR products revealed 62% mixed chromatograms predominantly belonging to Gram-positive bacteria. Terminal restriction fragment length polymorphism (T-RFLP) results of a sample subset confirmed sequence data showing polymicrobial DNA contents in 67% of bactDNA-positive ascites samples.
Sigma-Aldrich
CKI-7 dihydrochloride, ≥98% (HPLC), solid
Pricing and availability is not currently available.