Merck
  • The low FODMAP diet improves gastrointestinal symptoms in patients with irritable bowel syndrome: a prospective study.
The low FODMAP diet improves gastrointestinal symptoms in patients with irritable bowel syndrome: a prospective study.
International journal of clinical practice (2013-05-25)
R H de Roest, B R Dobbs, B A Chapman, B Batman, L A O'Brien, J A Leeper, C R Hebblethwaite, R B Gearry
ABSTRACT
Current treatment for irritable bowel syndrome (IBS) is suboptimal. Fermentable oligo-, di-, mono-saccharides and polyols (FODMAPs) may trigger gastrointestinal symptoms in IBS patients. Our aim was to determine whether a low FODMAP diet improves symptoms in IBS patients. Irritable bowel syndrome patients, who had performed hydrogen/methane breath testing for fructose and lactose malabsorption and had received dietary advice regarding the low FODMAP diet, were included. The effect of low FODMAP diet was prospectively evaluated using a symptom questionnaire. Furthermore, questions about adherence and satisfaction with symptom improvement, dietary advice and diet were assessed. Ninety patients with a mean follow up of 15.7 months were studied. Most symptoms including abdominal pain, bloating, flatulence and diarrhoea significantly improved (p < 0.001 for all). 75.6%, 37.8% and 13.3% of patients had fructose, lactose malabsorption or small intestinal bacterial overgrowth respectively. Fructose malabsorption was significantly associated with symptom improvement (abdominal pain odds ratio (OR) 7.09 [95% confidence interval (CI) 2.01-25.0], bloating OR 8.71 (95% CI 2.76-27.5), flatulence OR 7.64 (95% CI 2.53-23.0) and diarrhoea OR 3.39 (95% CI 1.17-9.78), p < 0.029 for all). Most patients (75.6%) were adherent to the diet, which was associated with symptom improvement (abdominal pain, bloating, flatulence and diarrhoea all significantly associated with adherence, r > 0.27, p < 0.011). Most patients (72.1%) were satisfied with their symptoms. The low FODMAP diet shows efficacy for IBS patients. The current strategy of breath testing and dietary advice provides a good basis to understand and adhere to the diet.
Product Description
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Pricing and availability is not currently available.
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.