Merck
  • Spectral studies on the interaction of pinacyanol chloride with binary surfactants in aqueous medium.
Spectral studies on the interaction of pinacyanol chloride with binary surfactants in aqueous medium.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2009-10-28)
Kausik Manna, Amiya Kumar Panda
ABSTRACT
Interaction of pinacyanol chloride (PIN) with pure and binary mixtures of cetyltrimethylammonium bromide (CTAB) and sodium deoxycholate (NaDC) was spectroscopically studied. Interaction of PIN with pure NaDC produced a blue shifted metachromatic band (at approximately 502 nm), which gradually shifted to higher wavelength region as the concentration of NaDC increased in the pre-micellar stage. For CTAB only intensity of both the bands increased without any shift. Mixed surfactant systems behaved differently than the pure components. Absorbance of monomeric band with a slight red-shift, and a simultaneous decrease in the absorbance of dimeric band of PIN, were observed for all the combinations in the post-micellar region. PIN-micelle binding constant (K(b)) for pure as well as mixed was determined from spectral data using Benesi-Hildebrand equation. Using the idea of Regular Solution Theory, micellar aggregates were assumed to be predominant than other aggregated state, like vesicles. Aggregation number was determined by fluorescence quenching method. Spectral analyses were also done to evaluate CMC values. Rubinigh's model for Regular Solution Theory was employed to evaluate the interaction parameters and micellar composition. Strong synergistic interaction between the oppositely charged surfactants was noted. Bulkier nature of NaDC lowered down its access in mixed micellar system.
Sigma-Aldrich
1,1′-Diethyl-2,2′-carbocyanine iodide, 97%
Sigma-Aldrich
Pinacyanol chloride