Merck
  • Enantioselective determination of sotalol enantiomers in biological fluids using high-performance liquid chromatography.
Enantioselective determination of sotalol enantiomers in biological fluids using high-performance liquid chromatography.
Journal of chromatography. B, Biomedical applications (1995-12-01)
T Shimizu, M Hiraoka, H Nakanomyo
ABSTRACT
A simple and sensitive high-performance liquid chromatographic (HPLC) method for the determination of (+)-(S)-sotalol and (-)-(R)-sotalol in biological fluids was established. Following extraction with isopropyl alcohol from biological samples on a Sep-Pak C18 cartridge, the eluent was derivatized with 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate (GITC). The diastereoisomeric derivates were resolved by HPLC with UV detection at 225 nm. Calibration was linear from 0.022 to 4.41 micrograms/ml in human plasma and from 0.22 to 88.2 micrograms/ml in human urine for both (+)-(S)- and (-)-(R)-sotalol. The lower limit of determination was 0.022 microgram/ml for plasma and 0.22 microgram/ml for urine. The within-day and day-to-day coefficients of variation were less than 7.5% for each enantiomer at 0.09 and 1.8 microgram/ml in plasma and at 0.44 and 4.4 micrograms/ml in urine. The method is also applicable to other biological specimens such as rat, mouse and rabbit plasma.
Product Description
Supelco
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate, for chiral derivatization, LiChropur, ≥98.0% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00
To order products, please contact your local dealer.