Merck
  • Internalization of Sambucus nigra agglutinins I and II in insect midgut CF-203 cells.
Internalization of Sambucus nigra agglutinins I and II in insect midgut CF-203 cells.
Archives of insect biochemistry and physiology (2011-01-22)
Shahnaz Shahidi-Noghabi, Els J M Van Damme, Winnok H De Vos, Guy Smagghe
ABSTRACT
In this project, the uptake mechanisms and localization of two lectins from Sambucus nigra, further referred to as S. nigra agglutinin (SNA)-I and SNA-II, into insect midgut CF-203 cells were studied. SNA-I is a chimeric lectin belonging to the class of ribosome-inactivating proteins, whereas SNA-II is a hololectin devoid of enzymatic activity. Internalization of the fluorescein isothiocyanate-labeled lectin was investigated using confocal microscopy. Both lectins were internalized into the cytoplasm of CF-203 cells at similar rates. Preexposure of the insect midgut cells to specific inhibitors of clathrin- and caveolae-mediated endocytosis resulted in an inhibition of lectin uptake in CF-203 cells and caspase-induced cytotoxicity caused by SNA-I and SNA-II, confirming the involvement of both endocytosis pathways. Further studies demonstrated that the uptake mechanism(s) for both lectins required phosphoinositide 3-kinases, but did not depend on the actin cytoskeleton. Since the hololectin SNA-II apparently uses a similar endocytosis pathway as the chimerolectin SNA-I, it can be concluded that the endocytosis process mainly relies on the carbohydrate-binding activity of the lectins under investigation. © 2011 Wiley Periodicals, Inc.
Sigma-Aldrich
Phenylarsine oxide, ≥97%, powder