Merck
  • Silica-based monolithic capillary columns-Effect of preparation temperature on separation efficiency.
Silica-based monolithic capillary columns-Effect of preparation temperature on separation efficiency.
Journal of chromatography. A (2010-08-03)
Josef Planeta, Dana Moravcová, Michal Roth, Pavel Karásek, Vladislav Kahle
ABSTRACT
The temperature effects during the sol-gel process and ageing of the silica-based monolith on the structure and separation efficiency of the capillary columns (100microm i.d., 150mm) for HPLC separations were studied. The tested columns were synthesized from a mixture of tetramethoxysilane, polyethylene glycol and urea under the acidic conditions. The temperature was varied from 40 degrees C to 44 degrees C and formation of bypass channels between the silica mold and the capillary wall was examined. The temperature of 43 degrees C was estimated as optimal for preparation of efficient silica capillary columns which were subsequently modified by octadecyldimethyl-N,N-diethylaminosilane or covered by poly(octadecyl methacrylate) and tested using standard mixture of alkylbenzenes under the isocratic conditions.
Sigma-Aldrich
Tetramethyl orthosilicate, 98%
Sigma-Aldrich
Tetramethyl orthosilicate, ≥99%
Sigma-Aldrich
Tetramethyl orthosilicate, purum, ≥98.0% (GC)