Merck
  • Development of a novel method based on liquid chromatography-evaporative light scattering detection for the direct determination of streptomycin and dihydrostreptomycin in raw materials, pharmaceutical formulations, culture media and plasma.
Development of a novel method based on liquid chromatography-evaporative light scattering detection for the direct determination of streptomycin and dihydrostreptomycin in raw materials, pharmaceutical formulations, culture media and plasma.
Journal of chromatography. A (2006-06-30)
Artemis K Sarri, Nikolaos C Megoulas, Michael A Koupparis
ABSTRACT
A novel method for the non-derivatization liquid chromatographic determination of streptomycin (STR) and dihydrostreptomycin (DHSTR) was developed and validated based on evaporative light scattering detection (ELSD). Utilizing a ThermoHypersil BetaBasic C18 analytical column, evaporation temperature of 50 degrees C and pressure of nebulizing gas (nitrogen) of 3.5 bar, the optimized mobile phase was 1.25 mL L(-1) TFA aqueous solution, in an isocratic mode at a rate of 1.0 mL min(-1). STR was eluted at 5.6 min and DHSTR at 7.8 min with a resolution of 4.4. Linear calibration curves were obtained from 2 to 120 microg mL(-1) (r > 0.9990) for STR and 2-75 microg mL(-1) (r > 0.9994) for DHSTR, with a LOD equal to 0.7 and 0.5 microg mL(-1), respectively. The developed method was applied for the assay of STR and DHSTR (sulfate) in pharmaceutical raw materials and formulations, while the simultaneous direct determination of sulfate was feasible (tR = 2.5 min, LOD = 1.4 microg mL(-1), double logarithmic calibration curve in the range of 4-50 microg mL(-1), r > 0.9998). Modified isocratic mobile phase (H2O-ACN, 90:10, v/v, containing 1.25 mL L(-1) TFA), was used for the determination of streptomycin B impurity in STR sulfate raw material and a gradient mobile phase (H2O-ACN containing TFA) was used for the determination of DHSTR in the presence of penicillinG procaine. The developed method was also applied for the assay of commercial formulations (STR powder and DHSTR injection solution and suspension) (%recovery 98-102, %RSD < 1.3, n = 3 x 3), for the determination of STR in bacteria culture medium (%recovery 99.6, %RSD = 0.8, n = 3 x 3), and for the determination of DHSTR in human plasma (2.0-23.0 microg mL(-1)) after solid phase extraction using carboxylate cartridges (%recovery 98.4-101.8, %RSD = 3.2, n = 3 x 3).
Sigma-Aldrich
Dihydrostreptomycin sesquisulfate, BioReagent, suitable for cell culture, ≥95.0% (HPLC)
Sigma-Aldrich
Dihydrostreptomycin sesquisulfate
Supelco
Dihydrostreptomycin Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Dihydrostreptomycin sulfate, European Pharmacopoeia (EP) Reference Standard