Merck
  • Endocrine Disruptor Bisphenol a Affects the Neurochemical Profile of Nerve Fibers in the Aortic Arch Wall in the Domestic Pig.
Endocrine Disruptor Bisphenol a Affects the Neurochemical Profile of Nerve Fibers in the Aortic Arch Wall in the Domestic Pig.
International journal of environmental research and public health (2022-05-29)
Liliana Rytel, László Könyves, Slawomir Gonkowski
ABSTRACT
Bisphenol A (BPA) is a synthetic compound utilized in industry for the production of various plastics. BPA penetrates into the environment and adversely affects living organisms. Therefore, the influence of various BPA dosages on the neurochemical characteristics of nerve fibers located in the aortic branch wall was investigated in this study utilizing a double immunofluorescence method. It was found that BPA in concentration of 0.5 mg/kg body weight/day causes a clear increase in the density of nerves within aortic branch walls immunoreactive to cocaine- and amphetamine-regulated transcript (CART), calcitonin gene-related peptide (CGRP), neuronal isoform of nitric oxide synthase (nNOS), pituitary adenylate cyclase-activating peptide (PACAP), and vasoactive intestinal polypeptide (VIP). Nerves containing galanin (GAL) and/or somatostatin (SOM) did not change when BPA was introduced into the system. Changes noted after administration of BPA at a dose of 0.05 mg/kg body weight/day were less visible and concerned fibers immunoreactive to CART, CGRP, and/or PACAP. The obtained results show that BPA affects the neurochemical coding of nerves in the aortic branch wall. These fluctuations may be the first signs of the influence of this substance on blood vessels and may also be at the root of the disturbances in the cardiovascular system.
Sigma-Aldrich
Anti-Nitric Oxide Synthase I Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-CARTPT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution