Merck
  • Proteogenomics analysis unveils a TFG-RET gene fusion and druggable targets in papillary thyroid carcinomas.
Proteogenomics analysis unveils a TFG-RET gene fusion and druggable targets in papillary thyroid carcinomas.
Nature communications (2020-04-30)
Aswini Krishnan, Jean Berthelet, Emilie Renaud, Sebastian Rosigkeit, Ute Distler, Eric Stawiski, Jing Wang, Zora Modrusan, Marc Fiedler, Mariann Bienz, Stefan Tenzer, Arno Schad, Wilfried Roth, Bernd Thiede, Somasekar Seshagiri, Thomas J Musholt, Krishnaraj Rajalingam
ABSTRACT
Papillary thyroid cancer (PTC) is the most common type of endocrine malignancy. By RNA-seq analysis, we identify a RET rearrangement in the tumour material of a patient who does not harbour any known RAS or BRAF mutations. This new gene fusion involves exons 1-4 from the 5' end of the Trk fused Gene (TFG) fused to the 3' end of RET tyrosine kinase leading to a TFG-RET fusion which transforms immortalized human thyroid cells in a kinase-dependent manner. TFG-RET oligomerises in a PB1 domain-dependent manner and oligomerisation of TFG-RET is required for oncogenic transformation. Quantitative proteomic analysis reveals the upregulation of E3 Ubiquitin ligase HUWE1 and DUBs like USP9X and UBP7 in both tumor and metastatic lesions, which is further confirmed in additional patients. Expression of TFG-RET leads to the upregulation of HUWE1 and inhibition of HUWE1 significantly reduces RET-mediated oncogenesis.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Roche
Cell Proliferation Kit I (MTT)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Triton X-100, laboratory grade
Millipore
ProteoExtract® Subcellular Proteome Extraction Kit
Sigma-Aldrich
BI8622, ≥98% (HPLC)
Sigma-Aldrich
BI8626, ≥98% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
Sigma-Aldrich
MBP, Dephosphorylated