Merck
  • P-glycoprotein deficiency enhances metabolic activation of and platelet response to clopidogrel through marked up-regulation of Cyp3a11 in mice: Direct evidence for the interplay between P-glycoprotein and Cyp3a.
P-glycoprotein deficiency enhances metabolic activation of and platelet response to clopidogrel through marked up-regulation of Cyp3a11 in mice: Direct evidence for the interplay between P-glycoprotein and Cyp3a.
Biochemical pharmacology (2020-11-03)
Jin-Zi Ji, Yi-Fei Li, Li-Ping Jiang, Ting Tai, Peng-Xin Ge, Qiong-Yu Mi, Ting Zhu, Hong-Guang Xie
ABSTRACT
Variability in P-glycoprotein (P-gp) efflux transporting activity was supposed to be involved in altered intestinal absorption and bioavailability of clopidogrel in patients; however, reliable evidence is still lacking. In this study, we sought to determine whether P-gp could play an important role in the metabolic activation of and platelet response to clopidogrel in mice. Abcb1a/1b knock-out (KO) and wild-type (WT) mice were used to evaluate differences in the intracellular accumulation of clopidogrel in the intestine, liver, and brain tissues and in systemic exposure of clopidogrel and its main metabolites as well as the mechanisms involved. Results indicated that, compared with WT mice, KO mice exhibited an 84% increase in systemic exposure of clopidogrel active thiol metabolite H4 and a 14.5% rise of suppression of ADP-induced platelet integrin αIIbβ3 activation, paralleled by a 41% decrease in systemic exposure of clopidogrel due to enhanced systemic clearance. Furthermore, KO mice displayed a 45% increase in Cyp3a11 but a 23% decrease in Ces1 at their protein levels compared with WT mice. Concurrently, intracellular clopidogrel concentrations in the tissues examined did not differ significantly between KO and WT mice. We conclude that although P-gp does not transport clopidogrel and its major metabolites in mice, P-gp-deficient mice exhibit elevated formation of the active metabolite H4 and enhanced antiplatelet effect of clopidogrel through up-regulation of Cyp3a11 and down-regulation of Ces1, suggesting that P-gp activity may correlate inversely with the formation of H4 and antiplatelet efficacy of clopidogrel in clinical settings due to P-gp and CYP3A4 interplay.
Sigma-Aldrich
2-Bromo-3′-methoxyacetophenone, 98%