Merck
  • GSAP regulates lipid homeostasis and mitochondrial function associated with Alzheimer's disease.
GSAP regulates lipid homeostasis and mitochondrial function associated with Alzheimer's disease.
The Journal of experimental medicine (2021-06-23)
Peng Xu, Jerry C Chang, Xiaopu Zhou, Wei Wang, Michael Bamkole, Eitan Wong, Karima Bettayeb, Lu-Lin Jiang, Timothy Huang, Wenjie Luo, Huaxi Xu, Angus C Nairn, Marc Flajolet, Nancy Y Ip, Yue-Ming Li, Paul Greengard
ABSTRACT
Biochemical, pathogenic, and human genetic data confirm that GSAP (γ-secretase activating protein), a selective γ-secretase modulatory protein, plays important roles in Alzheimer's disease (AD) and Down's syndrome. However, the molecular mechanism(s) underlying GSAP-dependent pathogenesis remains largely elusive. Here, through unbiased proteomics and single-nuclei RNAseq, we identified that GSAP regulates multiple biological pathways, including protein phosphorylation, trafficking, lipid metabolism, and mitochondrial function. We demonstrated that GSAP physically interacts with the Fe65-APP complex to regulate APP trafficking/partitioning. GSAP is enriched in the mitochondria-associated membrane (MAM) and regulates lipid homeostasis through the amyloidogenic processing of APP. GSAP deletion generates a lipid environment unfavorable for AD pathogenesis, leading to improved mitochondrial function and the rescue of cognitive deficits in an AD mouse model. Finally, we identified a novel GSAP single-nucleotide polymorphism that regulates its brain transcript level and is associated with an increased AD risk. Together, our findings indicate that GSAP impairs mitochondrial function through its MAM localization and that lowering GSAP expression reduces pathological effects associated with AD.
Sigma-Aldrich
Anti-PP1β Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)