Merck
  • Plasma cytokine and angiogenic factors associated with prognosis and therapeutic response to sunitinib vs everolimus in advanced non-clear cell renal cell carcinoma.
Plasma cytokine and angiogenic factors associated with prognosis and therapeutic response to sunitinib vs everolimus in advanced non-clear cell renal cell carcinoma.
Oncotarget (2017-02-09)
Pavlos Msaouel, Amado J Zurita, Shixia Huang, Eric Jonasch, Nizar M Tannir
ABSTRACT
No biomarkers are available to predict relative clinical benefit from targeted therapies in patients with non-clear cell renal cell carcinoma (nccRCC). To identify candidate predictive markers, we investigated a set of cytokines and angiogenic factors (CAFs) in previously untreated patients with nccRCC participating in the phase II ESPN trial comparing first-line sunitinib to everolimus. Pre-treatment concentrations of 30 CAFs were measured in plasma from 37 patients treated with everolimus (n=16) or sunitinib (n=21), and associated with progression-free (PFS) and overall survival (OS) after adjusting for potential confounders. High (>median) concentrations of soluble glycoprotein 130 (sgp130) were predictive of a longer PFS with sunitinib compared with everolimus (HR = 0.30; 95% CI: 0.11-0.85; P = 0.024). Significantly shorter PFS was noted, independently of treatment arm, in patients with high (>median) levels of IL-8 (HR = 3.13; 95% CI: 1.41-6.92), IL-13 (HR = 3.36; 95% CI: 1.49-7.58), and soluble tumor necrosis factor receptor II (HR = 2.21; 95% CI: 1.04-4.72). High IL-8 levels were also associated with significantly shorter OS (HR = 3.55; 95% CI: 1.55-8.14). Thus, using CAF profiling we identified candidate prognostic and predictive circulating biomarkers that can be used to inform therapeutic decisions in nccRCC.
Product Description
Millipore
MILLIPLEX® Human High Sensitivity T Cell Panel Premixed 13-plex - Immunology Multiplex Assay, Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.