Merck
  • Increased fibrotic signaling in a murine model for intra-arterial contrast-induced acute kidney injury.
Increased fibrotic signaling in a murine model for intra-arterial contrast-induced acute kidney injury.
American journal of physiology. Renal physiology (2020-03-24)
Amit Sharma, Sreenivasulu Kilari, Chuanqi Cai, Michael L Simeon, Sanjay Misra
ABSTRACT
Contrast-induced acute kidney injury (CI-AKI) is a vexing problem, and more than 70 million patients undergo studies using iodinated contrast. The molecular mechanisms responsible for CI-AKI are poorly understood. The goal of the present article was to determine the role of transforming growth factor-β1 (TGF-β1)/mothers against decapentaplegic homolog (SMAD)3 and associated collagen expression in a murine model of intra-arterial CI-AKI. The murine model of CI-AKI after intra-arterial contrast agent administration was created by first performing a partial nephrectomy to induce chronic kidney disease. Twenty-eight days later, 100 μL of contrast agent [iodixanol (320 mg/mL)] or saline were administered via the carotid artery. Two days after contrast administration, compared with saline, average serum creatinine was significantly elevated (P < 0.05). In the cortex, there was a significant increase in phosphorylated SMAD3 and gene expression of TGF-β1, TGF-β receptor type I, and TGF-β receptor type II at day 2 in the contrast group compared with the saline group. Average gene expressions of connective tissue growth factor, matrix metalloproteinase-2 and -9, and collagen type I-α and type IV-α were significantly increased at 2 days after contrast administration (all P < 0.05). Moreover, there was a decrease in Ki-67 staining in the cortex, with an increase in terminal deoxynucleotidyl transferase dUTP nick-end labeling in the cortex and medulla after contrast administration (P < 0.05). In the murine intra-arterial CI-AKI model, there was increased hypoxia and TGF-β1/SMAD3 pathway activation and collagen expression, resulting in renal fibrosis. Together, these results suggest that the TGF-β1/SMAD3 pathway could be a potential target in alleviating tissue fibrosis in CI-AKI.
Sigma-Aldrich
Anti-FSP1/S100A4 Antibody, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.