Merck
  • NH-sulfoximine: A novel pharmacological inhibitor of the mitochondrial F1 Fo -ATPase, which suppresses viability of cancerous cells.
NH-sulfoximine: A novel pharmacological inhibitor of the mitochondrial F1 Fo -ATPase, which suppresses viability of cancerous cells.
British journal of pharmacology (2020-10-11)
Daniela Strobbe, Rosalba Pecorari, Oriana Conte, Antonella Minutolo, Christine M M Hendriks, Stefan Wiezorek, Danilo Faccenda, Rosella Abeti, Carla Montesano, Carsten Bolm, Michelangelo Campanella
ABSTRACT
The mitochondrial F1 Fo -ATPsynthase is pivotal for cellular homeostasis. When respiration is perturbed, its mode of action everts becoming an F1 Fo -ATPase and therefore consuming rather producing ATP. Such a reversion is an obvious target for pharmacological intervention to counteract pathologies. Despite this, tools to selectively inhibit the phases of ATP hydrolysis without affecting the production of ATP remain scarce. Here, we report on a newly synthesised chemical, the NH-sulfoximine (NHS), which achieves such a selectivity. The chemical structure of the F1 Fo -ATPase inhibitor BTB-06584 was used as a template to synthesise NHS. We assessed its pharmacology in human neuroblastoma SH-SY5Y cells in which we profiled ATP levels, redox signalling, autophagy pathways and cellular viability. NHS was given alone or in combination with either the glucose analogue 2-deoxyglucose (2-DG) or the chemotherapeutic agent etoposide. NHS selectively blocks the consumption of ATP by mitochondria leading a subtle cytotoxicity associated via the concomitant engagement of autophagy which impairs cell viability. NHS achieves such a function independently of the F1 Fo -ATPase inhibitory factor 1 (IF1). The novel sulfoximine analogue of BTB-06584, NHS, acts as a selective pharmacological inhibitor of the mitochondrial F1 Fo -ATPase. NHS, by blocking the hydrolysis of ATP perturbs the bioenergetic homoeostasis of cancer cells, leading to a non-apoptotic type of cell death.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
D-Luciferin, synthetic
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Anti-AMBRA1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Ambra1 Antibody, from rabbit, purified by affinity chromatography