Merck
  • Glial Metabolic Rewiring Promotes Axon Regeneration and Functional Recovery in the Central Nervous System.
Glial Metabolic Rewiring Promotes Axon Regeneration and Functional Recovery in the Central Nervous System.
Cell metabolism (2020-09-18)
Feng Li, Armin Sami, Harun N Noristani, Kieran Slattery, Jingyun Qiu, Thomas Groves, Shuo Wang, Kelly Veerasammy, Yuki X Chen, Jorge Morales, Paula Haynes, Amita Sehgal, Ye He, Shuxin Li, Yuanquan Song
ABSTRACT
Axons in the mature central nervous system (CNS) fail to regenerate after axotomy, partly due to the inhibitory environment constituted by reactive glial cells producing astrocytic scars, chondroitin sulfate proteoglycans, and myelin debris. We investigated this inhibitory milieu, showing that it is reversible and depends on glial metabolic status. We show that glia can be reprogrammed to promote morphological and functional regeneration after CNS injury in Drosophila via increased glycolysis. This enhancement is mediated by the glia derived metabolites: L-lactate and L-2-hydroxyglutarate (L-2HG). Genetically/pharmacologically increasing or reducing their bioactivity promoted or impeded CNS axon regeneration. L-lactate and L-2HG from glia acted on neuronal metabotropic GABAB receptors to boost cAMP signaling. Local application of L-lactate to injured spinal cord promoted corticospinal tract axon regeneration, leading to behavioral recovery in adult mice. Our findings revealed a metabolic switch to circumvent the inhibition of glia while amplifying their beneficial effects for treating CNS injuries.
Sigma-Aldrich
(−)-Ethyl L-lactate, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
Sodium L-lactate, ≥99.0% (NT)
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Octyl-(S)-2HG, ≥98% (HPLC)