Merck
  • Abstracts and Case Studies From the College of American Pathologists 2018 Annual Meeting (CAP18).
Abstracts and Case Studies From the College of American Pathologists 2018 Annual Meeting (CAP18).
Archives of pathology & laboratory medicine (2018-09-13)
ABSTRACT
and case study poster sessions will be conducted during the 2018 College of American Pathologists Annual Meeting (CAP18), which is scheduled for October 20 to 24, 2018. The meeting will take place at the Hyatt Regency, Chicago, Illinois. The poster sessions will occur in the CAP18 Exhibit Hall. Specific dates and times for each poster session are listed below; "poster focus" times are dedicated poster viewing periods. Also shown before each poster session are the subject areas that will be presented.
Sigma-Aldrich
Anti-SV40 T Antigen (Ab-2) Mouse mAb (PAb416), liquid, clone PAb416, Calbiochem®
Pricing and availability is not currently available.