Merck
  • Aquaporin-3 expression in human fetal airway epithelial progenitor cells.
Aquaporin-3 expression in human fetal airway epithelial progenitor cells.
Stem cells (Dayton, Ohio) (2005-07-27)
Aurélie Avril-Delplanque, Ibrahim Casal, Nicolas Castillon, Jocelyne Hinnrasky, Edith Puchelle, Bruno Péault
ABSTRACT
Airway epithelium stem cells have not yet been prospectively identified, but it is generally assumed that both secretory and basal cells have the capacity to divide and differentiate. Previously, we developed a test for progenitor cells of the human airway epithelium, relying on the transplantation of fetal respiratory tissues into immunodeficient mice. In this study, we hypothesized that airway-repopulating epithelial progenitors can be marked with surface antigens, and we screened an array of such candidate markers, including lectin ligands, the CD44 and CD166 adhesion molecules, and the aquaporin-3 (AQP3) water channel. We observed that AQP3 is selectively expressed on the surface of basal cells, allowing the separation by flow cytometry of AQP3+ basal cells and AQP3- ciliated and secretory cells. Functional evaluation of sorted cells in vivo showed that AQP3+ cells can restore a normal pseudostratified, mucociliary epithelium as well as submucosal glands. AQP3- cells are also endowed with a similar potential, although faster engraftment suggests their inclusion of more committed progenitors. These results show that stem cell candidates in the human tracheo-bronchial mucosa can be positively selected with a novel marker but also, for the first time, that epithelial progenitors exist among both basal and suprabasal cell subsets within the human airway.
Sigma-Aldrich
Monoclonal Anti-Cytokeratin Peptide 13 antibody produced in mouse, clone KS-1A3, ascites fluid
Pricing and availability is not currently available.