Merck
  • Novel fibrillar structure in the inversin compartment of primary cilia revealed by 3D single-molecule superresolution microscopy.
Novel fibrillar structure in the inversin compartment of primary cilia revealed by 3D single-molecule superresolution microscopy.
Molecular biology of the cell (2020-01-03)
Henrietta W Bennett, Anna-Karin Gustavsson, Camille A Bayas, Petar N Petrov, Nancie Mooney, W E Moerner, Peter K Jackson
ABSTRACT
Primary cilia in many cell types contain a periaxonemal subcompartment called the inversin compartment. Four proteins have been found to assemble within the inversin compartment: INVS, ANKS6, NEK8, and NPHP3. The function of the inversin compartment is unknown, but it appears to be critical for normal development, including left-right asymmetry and renal tissue homeostasis. Here we combine superresolution imaging of human RPE1 cells, a classic model for studying primary cilia in vitro, with a genetic dissection of the protein-protein binding relationships that organize compartment assembly to develop a new structural model. We observe that INVS is the core structural determinant of a compartment composed of novel fibril-like substructures, which we identify here by three-dimensional single-molecule superresolution imaging. We find that NEK8 and ANKS6 depend on INVS for localization to these fibrillar assemblies and that ANKS6-NEK8 density within the compartment is regulated by NEK8. Together, NEK8 and ANKS6 are required downstream of INVS to localize and concentrate NPHP3 within the compartment. In the absence of these upstream components, NPHP3 is redistributed within cilia. These results provide a more detailed structure for the inversin compartment and introduce a new example of a membraneless compartment organized by protein-protein interactions.
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Pricing and availability is not currently available.
Millipore
Millex-HV Filter Unit (Sterile), pore size 0.45 μm, diam. 13 mm, PVDF membrane, hydrophilic, medical device
Pricing and availability is not currently available.