Merck
  • Low Serum Levels of (Dihydro-)Ceramides Reflect Liver Graft Dysfunction in a Real-World Cohort of Patients Post Liver Transplantation.
Low Serum Levels of (Dihydro-)Ceramides Reflect Liver Graft Dysfunction in a Real-World Cohort of Patients Post Liver Transplantation.
International journal of molecular sciences (2018-03-29)
Victoria Therese Mücke, Janis Gerharz, Katja Jakobi, Dominique Thomas, Nerea Ferreirós Bouzas, Marcus Maximilian Mücke, Sven Trötschler, Nina Weiler, Martin-Walter Welker, Stefan Zeuzem, Josef Pfeilschifter, Georgios Grammatikos
ABSTRACT
Patients after orthopic liver transplantation (OLT) are at risk of developing graft dysfunction. Sphingolipids (SL's) have been identified to play a pivotal role in the regulation of hepatocellular apoptosis, inflammation and immunity. We aimed to investigate the serum SL profile in a prospective real-world cohort of post-OLT patients. From October 2015 until July 2016, 149 well-characterized post-OLT patients were analyzed. SL's were assessed in serum probes via Liquid Chromatography/Tandem Mass Spectrometry. Twenty-nine (20%) patients had a biopsy proven graft rejection with decreased C20-ceramide (Cer) (p = 0.042), C18-dihydroceramide (DHC) (p = 0.022) and C24DHC (p = 0.060) levels. Furthermore, C18DHC (p = 0.044) and C24DHC (p = 0.011) were significantly down-regulated in patients with ischemic type biliary lesions (ITBL; n = 15; 10%). One-hundred and thirty-three patients (89%) have so far received tacrolimus as the main immunosuppressive agent with observed elevations of C14Cer (p = 0.052), C18Cer (p = 0.049) and C18:1Cer (p = 0.024). Hepatocellular carcinoma (HCC) pre-OLT was associated with increases in C24:1Cer (p = 0.024) and C24:1DHC (p = 0.024). In this large prospective cross-sectional study of patients, post-OLT serum levels of (very-)long chain (dihydro-)ceramides associate with graft rejection, ITBL, tacrolimus intake and HCC pre-OLT. Hence, serum SL's may be indicative of graft complications. Further research is necessary to identify their diverse mechanistic role in regulating immunity and inflammation in patients post-OLT.
Product Description
Avanti
C8 Dihydroceramide (d18:0/8:0), Avanti Research - A Croda Brand 860626P, powder
Avanti
C18 Dihydroceramide (d18:0/18:0), Avanti Research - A Croda Brand 860627P, powder
Avanti
C24:1 Dihydroceramide (d18:0/24:1(15Z)), Avanti Research - A Croda Brand 860629P, powder