Merck
  • Transmembrane chemokines act as receptors in a novel mechanism termed inverse signaling.
Transmembrane chemokines act as receptors in a novel mechanism termed inverse signaling.
eLife (2016-01-23)
Kirsten Hattermann, Henrike Gebhardt, Sebastian Krossa, Andreas Ludwig, Ralph Lucius, Janka Held-Feindt, Rolf Mentlein
ABSTRACT
The transmembrane chemokines CX3CL1/fractalkine and CXCL16 are widely expressed in different types of tumors, often without an appropriate expression of their classical receptors. We observed that receptor-negative cancer cells could be stimulated by the soluble chemokines. Searching for alternative receptors we detected that all cells expressing or transfected with transmembrane chemokine ligands bound the soluble chemokines with high affinity and responded by phosphorylation of intracellular kinases, enhanced proliferation and anti-apoptosis. This activity requires the intracellular domain and apparently the dimerization of the transmembrane chemokine ligand. Thus, shed soluble chemokines can generate auto- or paracrine signals by binding and activating their transmembrane forms. We term this novel mechanism "inverse signaling". We suppose that inverse signaling is an autocrine feedback and fine-tuning system in the communication between cells that in tumors supports stabilization and proliferation.
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Sigma-Aldrich
Anti-MAP Kinase 2/Erk2 Antibody, clone 1B3B9, clone 1B3B9, Upstate®, from mouse