Merck
  • Prevention of hepatocellular carcinoma by targeting MYCN-positive liver cancer stem cells with acyclic retinoid.
Prevention of hepatocellular carcinoma by targeting MYCN-positive liver cancer stem cells with acyclic retinoid.
Proceedings of the National Academy of Sciences of the United States of America (2018-04-25)
Xian-Yang Qin, Harukazu Suzuki, Masao Honda, Hikari Okada, Shuichi Kaneko, Ikuyo Inoue, Etsuko Ebisui, Kosuke Hashimoto, Piero Carninci, Keita Kanki, Hideki Tatsukawa, Naoto Ishibashi, Takahiro Masaki, Tomokazu Matsuura, Hiroyuki Kagechika, Kan Toriguchi, Etsuro Hatano, Yohei Shirakami, Goshi Shiota, Masahito Shimizu, Hisataka Moriwaki, Soichi Kojima
ABSTRACT
Hepatocellular carcinoma (HCC) is a highly lethal cancer that has a high rate of recurrence, in part because of cancer stem cell (CSC)-dependent field cancerization. Acyclic retinoid (ACR) is a synthetic vitamin A-like compound capable of preventing the recurrence of HCC. Here, we performed a genome-wide transcriptome screen and showed that ACR selectively suppressed the expression of MYCN, a member of the MYC family of basic helix-loop-helix-zipper transcription factors, in HCC cell cultures, animal models, and liver biopsies obtained from HCC patients. MYCN expression in human HCC was correlated positively with both CSC and Wnt/β-catenin signaling markers but negatively with mature hepatocyte markers. Functional analysis showed repressed cell-cycle progression, proliferation, and colony formation, activated caspase-8, and induced cell death in HCC cells following silencing of MYCN expression. High-content single-cell imaging analysis and flow cytometric analysis identified a MYCN+ CSC subpopulation in the heterogeneous HCC cell cultures and showed that these cells were selectively killed by ACR. Particularly, EpCAM+ cells isolated using a cell-sorting system showed increased MYCN expression and sensitivity to ACR compared with EpCAM- cells. In a long-term (>10 y) follow-up study of 102 patients with HCC, MYCN was expressed at higher levels in the HCC tumor region than in nontumor regions, and there was a positive correlation between MYCN expression and recurrence of de novo HCC but not metastatic HCC after curative treatment. In summary, these results suggest that MYCN serves as a prognostic biomarker and therapeutic target of ACR for liver CSCs in de novo HCC.
Sigma-Aldrich
Peretinoin, ≥97% (HPLC)