Merck
  • Spatially confined lignin nanospheres for biocatalytic ester synthesis in aqueous media.
Spatially confined lignin nanospheres for biocatalytic ester synthesis in aqueous media.
Nature communications (2018-06-14)
Mika Henrikki Sipponen, Muhammad Farooq, Jari Koivisto, Alessandro Pellis, Jani Seitsonen, Monika Österberg
ABSTRACT
Dehydration reactions proceed readily in water-filled biological cells. Development of biocatalysts that mimic such compartmentalized reactions has been cumbersome due to the lack of low-cost nanomaterials and associated technologies. Here we show that cationic lignin nanospheres function as activating anchors for hydrolases, and enable aqueous ester synthesis by forming spatially confined biocatalysts upon self-assembly and drying-driven aggregation in calcium alginate hydrogel. Spatially confined microbial cutinase and lipase retain 97% and 70% of their respective synthetic activities when the volume ratio of water to hexane increases from 1:1 to 9:1 in the reaction medium. The activity retention of industrially most frequently used acrylic resin-immobilized Candida antarctica lipase B is only 51% under similar test conditions. Overall, our findings enable fabrication of robust renewable biocatalysts for aqueous ester synthesis, and provide insight into the compartmentalization of diverse heterogeneous catalysts.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Amano Lipase M from Mucor javanicus, ≥10,000 U/g, pH 7.0, 40 °C (Optimum pH and temperature)
Pricing and availability is not currently available.