Merck
  • TRIM16 controls assembly and degradation of protein aggregates by modulating the p62-NRF2 axis and autophagy.
TRIM16 controls assembly and degradation of protein aggregates by modulating the p62-NRF2 axis and autophagy.
The EMBO journal (2018-08-26)
Kautilya Kumar Jena, Srinivasa Prasad Kolapalli, Subhash Mehto, Parej Nath, Biswajit Das, Pradyumna Kumar Sahoo, Abdul Ahad, Gulam Hussain Syed, Sunil K Raghav, Shantibhusan Senapati, Swati Chauhan, Santosh Chauhan
ABSTRACT
Sequestration of protein aggregates in inclusion bodies and their subsequent degradation prevents proteostasis imbalance, cytotoxicity, and proteinopathies. The underlying molecular mechanisms controlling the turnover of protein aggregates are mostly uncharacterized. Herein, we show that a TRIM family protein, TRIM16, governs the process of stress-induced biogenesis and degradation of protein aggregates. TRIM16 facilitates protein aggregate formation by positively regulating the p62-NRF2 axis. We show that TRIM16 is an integral part of the p62-KEAP1-NRF2 complex and utilizes multiple mechanisms for stabilizing NRF2. Under oxidative and proteotoxic stress conditions, TRIM16 activates ubiquitin pathway genes and p62 via NRF2, leading to ubiquitination of misfolded proteins and formation of protein aggregates. We further show that TRIM16 acts as a scaffold protein and, by interacting with p62, ULK1, ATG16L1, and LC3B, facilitates autophagic degradation of protein aggregates. Thus, TRIM16 streamlines the process of stress-induced aggregate clearance and protects cells against oxidative/proteotoxic stress-induced toxicity in vitro and in vivo Taken together, this work identifies a new mechanism of protein aggregate turnover, which could be relevant in protein aggregation-associated diseases such as neurodegeneration.
Sigma-Aldrich
MISSION® esiRNA, targeting human GABPA
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Mayer′s Hematoxylin Solution
Sigma-Aldrich
Scott′s Tap Water Substitute Concentrate, 10 ×