Merck
  • Reactive-site-centric chemoproteomics identifies a distinct class of deubiquitinase enzymes.
Reactive-site-centric chemoproteomics identifies a distinct class of deubiquitinase enzymes.
Nature communications (2018-03-23)
David S Hewings, Johanna Heideker, Taylur P Ma, Andrew P AhYoung, Farid El Oualid, Alessia Amore, Gregory T Costakes, Daniel Kirchhofer, Bradley Brasher, Thomas Pillow, Nataliya Popovych, Till Maurer, Carsten Schwerdtfeger, William F Forrest, Kebing Yu, John Flygare, Matthew Bogyo, Ingrid E Wertz
ABSTRACT
Activity-based probes (ABPs) are widely used to monitor the activity of enzyme families in biological systems. Inferring enzyme activity from probe reactivity requires that the probe reacts with the enzyme at its active site; however, probe-labeling sites are rarely verified. Here we present an enhanced chemoproteomic approach to evaluate the activity and probe reactivity of deubiquitinase enzymes, using bioorthogonally tagged ABPs and a sequential on-bead digestion protocol to enhance the identification of probe-labeling sites. We confirm probe labeling of deubiquitinase catalytic Cys residues and reveal unexpected labeling of deubiquitinases on non-catalytic Cys residues and of non-deubiquitinase proteins. In doing so, we identify ZUFSP (ZUP1) as a previously unannotated deubiquitinase with high selectivity toward cleaving K63-linked chains. ZUFSP interacts with and modulates ubiquitination of the replication protein A (RPA) complex. Our reactive-site-centric chemoproteomics method is broadly applicable for identifying the reaction sites of covalent molecules, which may expand our understanding of enzymatic mechanisms.
Sigma-Aldrich
Anti-RPA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ZUFSP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution