Merck
  • Capzimin is a potent and specific inhibitor of proteasome isopeptidase Rpn11.
Capzimin is a potent and specific inhibitor of proteasome isopeptidase Rpn11.
Nature chemical biology (2017-03-01)
Jing Li, Tanya Yakushi, Francesco Parlati, Andrew L Mackinnon, Christian Perez, Yuyong Ma, Kyle P Carter, Sharon Colayco, Gavin Magnuson, Brock Brown, Kevin Nguyen, Stefan Vasile, Eigo Suyama, Layton H Smith, Eduard Sergienko, Anthony B Pinkerton, Thomas D Y Chung, Amy E Palmer, Ian Pass, Sonja Hess, Seth M Cohen, Raymond J Deshaies
ABSTRACT
The proteasome is a vital cellular machine that maintains protein homeostasis, which is of particular importance in multiple myeloma and possibly other cancers. Targeting of proteasome 20S peptidase activity with bortezomib and carfilzomib has been widely used to treat myeloma. However, not all patients respond to these compounds, and those who do eventually suffer relapse. Therefore, there is an urgent and unmet need to develop new drugs that target proteostasis through different mechanisms. We identified quinoline-8-thiol (8TQ) as a first-in-class inhibitor of the proteasome 19S subunit Rpn11. A derivative of 8TQ, capzimin, shows >5-fold selectivity for Rpn11 over the related JAMM proteases and >2 logs selectivity over several other metalloenzymes. Capzimin stabilized proteasome substrates, induced an unfolded protein response, and blocked proliferation of cancer cells, including those resistant to bortezomib. Proteomic analysis revealed that capzimin stabilized a subset of polyubiquitinated substrates. Identification of capzimin offers an alternative path to develop proteasome inhibitors for cancer therapy.
Sigma-Aldrich
Capzimin dimer, ≥96% (HPLC)