Xanthurenic acid
Synonym(s):
4,8-Dihydroxyquinaldic acid, 4,8-Dihydroxyquinoline-2-carboxylic acid
Empirical Formula (Hill Notation):
C10H7NO4
CAS Number:
Molecular Weight:
205.17
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Showing 1-1 of 1