
Thioridazine hydrochloride

Synonym(s):
10-[2-(1-Methyl-2-piperidyl)ethyl]-2-(methylthio)-10H-phenothiazine hydrochloride, Thioridazine hydrochloride
Empirical Formula (Hill Notation):
C21H26N2S2 · HCl
CAS Number:
Molecular Weight:
407.04
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing