Skip to Content
Merck
Sodium cyanoborohydride solution 1.0 M in THF

Sodium cyanoborohydride solution

Sodium cyanoborohydride solution 1.0 M in THF
Synonym(s):
Sodium borocyanohydride, Sodium cyanoborohydride, Sodium cyanotrihydroborate
Linear Formula:
NaBH3CN
CAS Number:
Molecular Weight:
62.84

Products

Product Category
Brand
Formula Weight
Quality Segment
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing