
Rizatriptan for system suitability

Synonym(s):
Rizatriptan benzoate salt, MK-462, N,N-Dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethylamine benzoate salt
Empirical Formula (Hill Notation):
C15H19N5 · C7H6O2
CAS Number:
Molecular Weight:
391.47
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing